Order Entry
ContactUsLinkComponent
Chiral ART Amylose-SA Columns, YMC America
Chiral ART Amylose-SA Columns, YMC America
Catalog #: 10498-460
Supplier:  YMC America,Inc
Chiral ART Amylose-SA Columns, YMC America
Catalog #: 10498-460
Supplier:  YMC America,Inc

Specifications

  • I.D.:
    4.60 mm
  • Length:
    250 mm
  • Cat. no.:
    10498-460
  • Packing Material:
    Silica
  • Phase:
    Amylose-SA
  • Supplier No.:
    KSA99S052546WT

Specifications

About this item

CHIRAL ART Polysaccharide Derivative Series for cost-effective, reproducible Chiral separations.

  • Applicability to various chiral compounds
  • Novel high-strength
  • Super-wide-pore silica gel base
  • High durability that is suitable for SFC/SMB

CHIRAL ART Polysaccharide Derivative Series for cost-effective, reproducible Chiral separations. It include a Chiral HPLC column/packing material with polysaccharides chiral selector.

Customers Who Bought This Also Bought