Nebulizer Cleaning Wires, PerkinElmer
Catalog #: CA97013-702
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:Nebulizer Cleaning Wires
- Cat. no.:CA97013-702
- Supplier No.:03030135
Specifications
About this item
Made of soft alloy copper, these cleaning wires are specially deburred to prevent scratching of the orifices and internal components of the nebulizers while cleaning.
- Soft copper construction
- Deburred
- Provided in convenient packs of multiples
Use only PerkinElmer cleaning wires to clean nebulizers.
Customers Who Bought This Also Bought