Specifications
- Well Volume:100 µL
- Description:Nunc™ MaxiSorp™ ELISA Plates
- No. of wells:96
- Cat. no.:CA10761-500
- Supplier No.:423501
Specifications
About this item
Flat-bottom 96-well plates feature a hydrophilic/hydrophobic mixed surface that binds to a wide variety of biomolecules
- Made from a highly controlled and characterized resin
- Uncoated plates
Used with BioLegend ELISA MAX™ sets as well as ELISA antibody pairs.
This makes it an ideal choice to bind most antigens or primary antibodies during ELISA.