Corning® CoolBox™ XT All-Day Cooling and Freezing Workstations, Corning
Supplier: Corning
Certificates
About this item
Ideal for transfering samples between labs, or in the field.
- Over 16 hours of 0,5 to 4 °C cooling with lid closed and over 10 hours with the lid open
- Reduces contamination by eliminating ice
- Easy to clean with lab detergents or bleach
- Patent-pending dual phase conductive XT Cooling core ensures stable and consistent sample cooling
- Portable XT CoolBox systems can be used when a brief transfer of temperature sensitive samples to a robotic device is needed
- May be used with dry ice in place of the Cooling Core to create a portable snap-freezing workstation
CoolBox™ XT ice-free bench top systems provide sample cooling or freezing without ice, electricity or batteries.
CoolBox XT workstations cool via thermoconductivity. Place the CoolRack module on top of the pre-frozen cooling core, included in the CoolBox XT and the samples achieve 0 °C in less than three minutes.
Design your own CoolBox system. CoolBox XT includes one cooling core and accomodates one sample module. CoolBox 2XT includes 2 cooling cores and accomodates two sample modules. Order CoolRack modules separately based on tube or plate size required.
Ordering information: See accessories for additional cooling cores as well as an optional freezing core for sample processing below 0 °C. Purchase CoolRack sample modules separately.
Related Information
Certificates
Search by Lot Number3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture Quick Reference Ordering Guide Corning® CoolBox™ and CoolRack® Consumable Compatibility Guide Corning® CoolBox™ Ice-free Systems Corning® CoolBox™ System for Consistent and Reproducible Cooling
Product Literature
More
75779-864_26218019
Conformity declaration
Corning New Lab Startup Program
Promotion Material
Customers Who Bought This Also Bought