Order Entry
Canada
ContactUsLinkComponent
Single-Stage Ultra-High Purity Stainless Steel Gas Regulators with International Fittings, Restek
Single-Stage Ultra-High Purity Stainless Steel Gas Regulators with International Fittings, Restek
Supplier:  Restek
Certificates
undefined
Single-Stage Ultra-High Purity Stainless Steel Gas Regulators with International Fittings, Restek
Supplier:  Restek

Specifications

  • Catalog No:
  • 10854-498
  • 10855-468
  • 10854-500
  • 10855-464
  • 10855-466
  • Outlet gauge:
  • 30", 0 to 14 bar (0 – 200 psig)
  • 30", 0 to 14 bar (0 – 200 psig)
  • 30", 0 to 14 bar (0 – 200 psig)
  • 30", 0 to 14 bar (0 – 200 psig)
  • 30", 0 to 14 bar (0 – 200 psig)

Specifications

Frequently Bought Together