Dual-Stage Ultra-High Purity Stainless Steel Gas Regulators with International Fittings, Restek
Supplier: Avantor
Certificates
About this item
UHP stainless steel regulators are the standard for ultra-high-purity and corrosion-resistant pressure regulation.
- Secondary pressure regulation not needed
- Most stable outlet pressure control
- Also available with CGA fittings
All regulators are rated to 3000 psig maximum inlet pressure.
Specifications
- Catalog No:
- 10855-580
- 10855-538
- 10855-582
- Outlet gauge:
- 30", 0 to 14 bar (0 – 200 psig)
- 30", 0 to 14 bar (0 – 200 psig)
- 30", 0 to 14 bar (0 – 200 psig)