Double Taper Inlet Liner for PerkinElmer Auto SYS™ and Clarus GCs equipped with split/splitless inlets, Restek
Supplier: Restek
Certificates
About this item
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Customers Who Bought This Also Bought