Metric 9-Piece, Ball-Point Hex Key Set, Restek
Catalog #: 10058-834
Supplier: Restek
Specifications
- Description:Metric 9-Piece, Ball-Point Hex Key Set
- Cat. no.:10058-834
- Supplier No.:22999
Specifications
About this item
Ball-point hex key style includes nine metric hex keys (Allen wrenches): 1.5, 2, 2.5, 3, 4, 5, 6, 8, and 10 mm.
Customers Who Bought This Also Bought