Gold Tip Transfer Line, Restek
Catalog #: 10856-138
Supplier: Restek
Specifications
- Description:Gold Tip Transfer Line
- Cat. no.:10856-138
- Supplier No.:24699
Specifications
About this item
For use with Agilent 5971/5972 MS systems.
- Gold-plated for inertness
- Meets or exceeds original manufacturer's performance
Customers Who Bought This Also Bought