Spanner Wrench for Agilent 5890/6850/6890/7890 FID Collector Assemblies, Restek
Catalog #: 10852-848
Supplier: Restek
Specifications
- Description:Spanner Wrench
- Cat. no.:10852-848
- For Use With:for Agilent 5890/6850/6890/7890 FID Collector Assembly
- Supplier No.:22329
Specifications
About this item
Use a spanner wrench to easily remove a nut from an FID collector without damaging the nut.
- Unique, ergonomic handle that is easy to grip
- Fits all instrument models
Customers Who Bought This Also Bought