Inlet Wrench for Shimadzu 17A, 2010, and 2014 GCs, Restek
Catalog #: 10852-852
Supplier: Restek
Specifications
- Description:Inlet Wrench
- Cat. no.:10852-852
- Supplier No.:21159
Specifications
About this item
For use with Shimadzu 17A, 2010, and 2014 GCs.
- Designed specifically for removing Shimadzu injection ports
- High-quality stainless steel construction
Customers Who Bought This Also Bought