Single-Stage Ultra-High Purity Chrome-Plated Brass Gas Regulators with CGA Fittings, Restek
Supplier: Restek
Certificates
About this item
Use this ultra-high-purity gas regulator when there is secondary pressure regulation downstream.
- Identical gas purity protection as with dual-stage gas regulators
- Oxidation-resistant
- Chrome-plated
- Also available with international fittings
Brass regulators feature reduced internal dead volume relative to stainless steel bodies.
Specifications
- Catalog No:
- 10854-502
- 10855-238
- 10855-542
- Outlet gauge:
- 30", 0 to 200 psig (0 – 1379 kPa)
- 30", 0 to 200 psig (0 – 1379 kPa)
- 30", 0 to 200 psig (0 – 1379 kPa)