Scoring Wafer with Handle, Restek
Catalog #: 97040-464
Supplier: Restek
Specifications
- Description:Scoring Wafer with Handle
- Cat. no.:97040-464
- Supplier No.:23015
Specifications
About this item
Ceramic scoring wafer is straight-edged for cutting fused silica tubing cleanly.
- Unique, ergonomic handle is made of soft, comfortable rubber
Customers Who Bought This Also Bought