ContactUsLinkComponent
Universal Cylinder Wrench, Restek
Universal Cylinder Wrench, Restek
Catalog #: 10855-804
Supplier:  Restek
Universal Cylinder Wrench
Universal Cylinder Wrench, Restek
Catalog #: 10855-804
Supplier:  Restek
Specifications
  • Description:
    Universal Cylinder Wrench
  • Cat. no.:
    10855-804
  • Supplier No.:
    21322
Specifications
About this item
Use this versatile wrench for tightening gauges and gas regulator CGA fittings to cylinder outlets and pipe thread connections.
  • Multiple sizes allow for universal matching with any system
Customers Who Bought This Also Bought