ContactUsLinkComponent
Cylinder Valve Wrench, Restek
Cylinder Valve Wrench, Restek
Catalog #: 10855-832
Supplier:  Restek
Cylinder Valve Wrench
Cylinder Valve Wrench, Restek
Catalog #: 10855-832
Supplier:  Restek
Specifications
  • Description:
    Cylinder Valve Wrench
  • Cat. no.:
    10855-832
  • Supplier No.:
    21321
Specifications
About this item
This specially designed wrench enables easy opening of cylinder valves that are fitted with a hand wheel.
It is also suitable for removing difficult cylinder caps.
Customers Who Bought This Also Bought