Dual-Stage Ultra-High Purity Chrome-Plated Brass Gas Regulators with CGA Fittings, Restek
Supplier: Restek
Certificates
About this item
Oxidation-resistant, chrome-plated dual-stage brass gas regulators are ideal for sensitive GC applications using MS, PID, or ECD detection methods.
- Most stable outlet pressure control
- Secondary pressure regulation not needed
- Most widely used regulator
- Less internal volume than stainless steel gas regulators
Regulators feature CGA fittings, and are also available with international fittings. Custom gas regulators are also available.
Specifications
- Catalog No:
- RK21668
- RK21667
- RK21669
- Outlet pressure:
- 0–100 psig
- 0–100 psig
- 0–100 psig
- Inlet gauge:
- 0–4000 psig
- 0–4000 psig
- 0–4000 psig
- Outlet gauge:
- 30", 0–200 psig
- 30", 0–200 psig
- 30", 0–200 psig
- Outlet type:
- Diaphragm Valve, 1/4" Tube Fitting
- Diaphragm Valve, 1/4" Tube Fitting
- Diaphragm Valve, 1/4" Tube Fitting