Parker HydroGen Mate™ DI Water System, Restek
Catalog #: RK22000
Supplier: Restek
Specifications
- Description:HydroGen Mate DI Water System-Model 72-230
- Cat. no.:RK22000
- Supplier No.:22000
Specifications
About this item
Economical means of providing deionized water to hydrogen generators, or for other uses.
- Quick and easy installation with minimal maintenance
- Removes organics, phosphates, chlorine, and essentially all ionizable constituents from your water supply
- Easy-fill dispensing gun
- Visual indication for cartridge changes
This system is specifically designed to provide the high-purity deionized water needed by all models of Parker hydrogen generators. The system is ready to install and is shipped complete with prefiltration cartridge, two DI resin exchange cartridges, final filter, and dispensing gun. Maintenance Kit includes two resin exchange cartridges, 1 prefilter, and 1 final filter.
Customers Who Bought This Also Bought