Purge-and-Trap Spargers, Restek
Supplier: Restek
Certificates
About this item
Available with uniform frits to ensure maximum purging efficiency.
- Use non-fritted spargers for wastewater samples
- Manufactured to tight tolerances to ensure a leak-tight seal
- Designed for use with Tekmar purge-and-trap instruments only
Customers Who Bought This Also Bought