Packed Column FID Replacement Jets for Agilent 5890/6850/6890 GCs, Restek
Supplier: Restek
Certificates
About this item
Available in standard or Siltek®-treated high-performance versions.
- Threads specially coated for easy installation and removal
- Special processing ensures the highest degree of cleanliness
- Meets or exceeds original manufacturer's performance
- Can be Siltek® treated to be extremely inert, for use with active compounds
For use with Agilent 5890/6850/6890 GCs
0.018" ID jets: Used for most general-purpose packed column applications.
0.030" ID jets: For packings that exhibit high bleed and that frequently clog the tip of smaller 0.018" jets.
Customers Who Bought This Also Bought