Ceramic Scoring Wafer, Restek
Catalog #: RK-20116
Supplier: Restek
Specifications
- Description:Ceramic Scoring Wafers
- Cat. no.:RK-20116
- Supplier No.:20116
Specifications
About this item
Four straight scoring edges for cutting fused silica tubing and four serrated edges for cutting MXT metal capillary columns.
- Marked increments for easily measured scoring
Crafted from durable ceramic material.
Customers Who Bought This Also Bought