Sapphire Scribe, Restek
Catalog #: 10852-902
Supplier: Restek
Specifications
- Description:Sapphire Scribe
- Cat. no.:10852-902
- Supplier No.:20182
Specifications
About this item
Sapphire scribe allows the user to cut fused silica tubing cleanly and in a square format.
Customers Who Bought This Also Bought