Specifications
- Description:Quickslide™ gram stain control slides
- Cat. no.:75779-644
- Supplier No.:AGS-00700
Specifications
About this item
GramPRO 1™ Gram stain control slides are prepared from standard stock ATCC quality control organisms
The Gram positive organism is Staphylococcus aureus. The Gram negative organism is E.coli. They are intended to be used in conjunction with the GramPRO 1™ Automatic Gram Stain Instrument, and may also be used for manual staining. Slides are provided in packs of 50.