Temperature Probe for Sonifier® SFX and 450D Series, Branson Ultrasonics
Catalog #: 75795-830
Supplier: Branson Ultrasonics
Specifications
- Description:Temperature Probe for SFX and 450D Sonifiers®
- Cat. no.:75795-830
- Supplier No.:200-060-022R
Specifications
About this item
The Temperature Probe accessory is a must-have for monitoring the temperature of sensitive samples
With Branson’s industry-exclusive True Temperature Control, the SFX Sonifiers® will automatically adjust the ultrasonic pulse length to maintain a temperature within a set range. Simply set a Pulse Temperature (the lower limit of the desired temperature range) and a Max. Temperature (upper limit), and the Sonifier® does the rest.
Simply plug the probe into the back of the Digital Sonifier®, and position its tip at the desired depth within a sample.
Customers Who Bought This Also Bought