Ocean Pacific® Eclipse™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Supplier: Medicom
Certificates
About this item
Lightweight nitrile glove with a wider palm.
- Lightweight design provides exceptional comfort through the palm and thumb
- Additional thickness at fingertips ensures durability in the most crucial areas
- Internal surface is polymer coated for improved donning and biocompatibility
- Textured fingertips and bonded polymer exterior surface ensure a secure grip, even when wet
Ultra-light for optimal dexterity and tactility. Gloves conform to hands to help minimize fatigue and maximize tactility. Textured fingertips ensure a secure grip, even when wet.
Non sterile, single-use, medical examination gloves are intended to prevent transmission of a wide variety of diseases to both patients and healthcare personnel. Specifically, gloves are intended to act as a barrier between the healthcare provider and patients to prevent contamination between patient and caregiver
Certifications: ASTM D6978-05 certified for chemotherapy handling.
Packaging: Gloves are packaged 200/box, 10 boxes/case.
Specifications
- Catalog No:
- CA75795-654
- CA75795-656
- CA75795-706
- CA75795-708
- CA75795-710
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Finger thickness:
- 0.05 mm
- 0.05 mm
- 0.05 mm
- 0.05 mm
- 0.05 mm
- Palm thickness:
- 0.05 mm
- 0.05 mm
- 0.05 mm
- 0.05 mm
- 0.05 mm