Order Entry
ContactUsLinkComponent
Human Recombinant IL-27 non-lytic (from CHO Cells)
Human Recombinant IL-27 non-lytic (from CHO Cells)
Catalog #: 75794-420
Supplier:  Prosci
CAS Number:  
Human Recombinant IL-27 non-lytic (from CHO Cells)
Catalog #: 75794-420
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    50 μg
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Biological activity:
    Bioactivity was measured in a cell proliferation assay using anti-CD3 activated human peripheral mononuclear cells.
  • Protein synonyms:
    Interleukin-27
  • Protein/peptide name:
    IL-27 non-lytic
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    26-243
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.
  • Cat. no.:
    75794-420
  • Supplier No.:
    90-529

Specifications

About this item

Interleukin-27 (IL-27) is a heterodimeric group 2 receptor ligand molecule that belongs to the IL-6/IL-12 family of long type I cytokines. It is composed of EBI3 (EBV-induced gene 3), a 34 kDa glycoprotein that is related to the p40 subunit of IL-12 and IL-23, and p28, the cloned 28 kDa glycoprotein that is related to the p35 chain of IL-12. IL-27 is expressed by monocytes, endothelial cells and dendritic cells. IL-27 binds to and signals through a heterodimeric receptor complex composed of WSX1 (TCCR) and gp130. Evidence suggests IL-27 interacts only with WSX-1. IL-27 has both anti- and proinflammatory properties. As an antiinflammatory, IL-27 seems to induce a general negative feedback program that limits T and NK-T cell activity. At the onset of infection, IL-27 induces an IL-12 receptor on naie CD4+ T cells, making them susceptible to subsequent IL-12 activity (and possible Th1 development). Notably, IL-12 family cytokines are both induced and inhibited by bacterial products. Microbes promote IL-27 secretion through TLR4, and also block IL-27 production via C5a induction.

  • Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.

Customers Who Bought This Also Bought