Order Entry
ContactUsLinkComponent
H. influenzae Recombinant NAD Nucleotidase (from E. coli), ProSci
H. influenzae Recombinant NAD Nucleotidase (from E. coli), ProSci
Catalog #: 75794-330
Supplier:  Prosci
CAS Number:  
H. influenzae Recombinant NAD Nucleotidase (from E. coli), ProSci
Catalog #: 75794-330
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    H. influenzae
  • Size:
    50 μg
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at –80 °C.
  • Enzyme name:
    NAD Nucleotidase
  • Enzyme activity:
    With AMP: 856.4 nmol of Ade/min/mg of protein
    With NMN: 110,3 nmol of NR/min/mg of protein
    With NAD: 5.4 nmol of Ade/min per mg of protein | 5.3 nmol of NR/min/mg of protein
  • Enzyme synonyms:
    NadN, NAD Pyrophosphatase/5'-nucleotidase
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    Full-length NAD 5’-nucleotidase without the signal peptide corresponding to the first 26 aa
  • Formulation:
    Liquid, In 5mM KH₂PO₄, pH 6.8 containing 100 mM TRIS-HCl (pH 8.5) and 10 mM MgCl₂.
  • Cat. no.:
    75794-330
  • Supplier No.:
    90-368

Specifications

About this item

NadN (NAD nucleotidase) is a periplasmic enzyme from Haemophilus influenzae, a major pathogen of the respiratory tract in humans that has developed the capability to exploit host NAD(P) for its nicotinamide dinucleotide requirement

This recombinant proteins is for research use only.

Fusion-Tag: Tag Free

NadN plays a central role by degrading NAD into adenosine and NR (nicotinamide riboside), NMN to nicotinamide riboside and AMP to adenosine. It shows a broad substrate specificity, recognizing either mono- or di-nucleotide nicotinamides and different adenosine phosphates with a maximal activity on 5'-adenosine monophosphate. It can be used as an economically alternative to produce NR (nicotinamide riboside). Catalytic activity: NAD=AMP + NMN=Ade+NR+PPi | AMP=Ade+Pi | NMN=NR+Pi

Customers Who Bought This Also Bought