Order Entry
ContactUsLinkComponent
Human Recombinant TIP60 (highly active) (from Insect Cells (Sf21))
Human Recombinant TIP60 (highly active) (from Insect Cells (Sf21))
Catalog #: 75794-160
Supplier:  Prosci
CAS Number:  
Human Recombinant TIP60 (highly active) (from Insect Cells (Sf21))
Catalog #: 75794-160
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf21)
  • Species:
    Human
  • Size:
    2 μg
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at −80 °C.
  • Biological activity:
    100-200ng are required for standard HAT assay.
  • Protein synonyms:
    Histone Acetyltransferase HTATIP|Lysine Acetyltransferase 5|Histone Acetyltransferase KAT5|60 kDa Tat-interactive Protein
  • Protein/peptide name:
    TIP60 (highly active)
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Liquid, In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride, 0.2% NP-40, 50mM imidazole and 10% glycerol.
  • Cat. no.:
    75794-160
  • Supplier No.:
    90-347

Specifications

About this item

TIP60 is the catalytic subunit of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of selected genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome-DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex is required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis and DNA repair. NuA4 may also play a direct role in DNA repair when recruited to sites of DNA damage. TIP60 directly acetylates and activates ataxia-telangiectasia mutated (ATM). In case of HIV-1 infection, interaction with the viral Tat protein leads to KAT5 polyubiquitination and targets it to degradation.

  • Fusion-Tag: His Tag

This recombinant proteins is for research use only.

Customers Who Bought This Also Bought