Order Entry
ContactUsLinkComponent
Human Recombinant GCN5 Highly Active (from Insect Cells (Sf21))
Human Recombinant GCN5 Highly Active (from Insect Cells (Sf21))
Catalog #: 75794-156
Supplier:  Prosci
CAS Number:  
Human Recombinant GCN5 Highly Active (from Insect Cells (Sf21))
Catalog #: 75794-156
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf21)
  • Species:
    Human
  • Size:
    2 μg
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at −80 °C.
  • Biological activity:
    ~200-300ng are required for standard HAT assay.
  • Protein synonyms:
    Lysine Acetyltransferase 2A|STAF97|GCN5L2|Histone Acetyltransferase KAT2A|Histone Acetyltransferase GCN5
  • Protein/peptide name:
    GCN5 highly active
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Liquid, In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride, 0.2% NP-40, 50mM imidazole and 10% glycerol.
  • Cat. no.:
    75794-156
  • Supplier No.:
    90-345

Specifications

About this item

GCN5 functions as a histone acetyltransferase (HAT) to promote transcriptional activation. Acetylation of histones gives a specific tag for epigenetic transcription activation. It has significant histone acetyltransferase activity with core histones, but not with nucleosome core particles. In case of HIV-1 infection, it is recruited by the viral protein Tat. It regulates Tat's transactivating activity and may help inducing chromatin remodeling of proviral genes. GCN5 is a component of the Ada Two-A containing (ATAC) complex, a complex with histone acetyltransferase activity on histones H3 and H4.

  • Fusion-Tag: His Tag

This recombinant proteins is for research use only.

Customers Who Bought This Also Bought