Order Entry
ContactUsLinkComponent
Mouse Recombinant CREB-binding highly active (from Insect cells (Sf21))
Mouse Recombinant CREB-binding highly active (from Insect cells (Sf21))
Catalog #: 75794-154
Supplier:  Prosci
CAS Number:  
Mouse Recombinant CREB-binding highly active (from Insect cells (Sf21))
Catalog #: 75794-154
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf21)
  • Species:
    Mouse
  • Size:
    2 μg
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at −80 °C.
  • Biological activity:
    ~50-100ng are required for standard HAT assay.
  • Protein synonyms:
    CBP|CREBBP
  • Protein/peptide name:
    CREB-binding highly active
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Liquid, In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride, 0.2% NP-40, 50-100mM imidazole and 10% glycerol.
  • Cat. no.:
    75794-154
  • Supplier No.:
    90-344

Specifications

About this item

CREB-binding protein acetylates histones, giving a specific tag for transcriptional activation. It also acetylates non-histone proteins, like NCOA3 and FOXO1. It binds specifically to phosphorylated CREB and enhances its transcriptional activity toward cAMP-responsive genes. Acts as a coactivator of ALX1 in the presence of EP300.

  • Fusion-Tag: His Tag

This recombinant proteins is for research use only.

Customers Who Bought This Also Bought