To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:10 μg
- Tag sequence:His Tag
- Storage conditions:Working aliquots are stable for up to 3 months when stored at −20 °C.
- Protein synonyms:FATSO|Fat Mass and Obesity-associated Protein
- Protein/peptide name:FTO
- Purity:>90% (SDS-PAGE)
- Sequence:2-505
- Formulation:0.2um-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl.
- Cat. no.:75793-704
- Supplier No.:90-119
Specifications
About this item
FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.
- Fusion-Tag: His Tag
This recombinant proteins is for research use only.