Order Entry
ContactUsLinkComponent
Rat Recombinant NAMPT (from HEK293 Cells)
Rat Recombinant NAMPT (from HEK293 Cells)
Catalog #: 75793-612
Supplier:  Prosci
CAS Number:  
Rat Recombinant NAMPT (from HEK293 Cells)
Catalog #: 75793-612
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Size:
    50 μg
  • Tag sequence:
    DDDDK Tag
  • Storage conditions:
    Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    Pre-B Cell Colony Enhancing Factor 1|Nicotinamide Phosphoribosyltransferase|PBEF1
  • Protein/peptide name:
    NAMPT
  • Purity:
    >90% (SDS-PAGE)
  • Sequence:
    1-491
  • Formulation:
    0.2um-filtered solution in PBS, pH 7.2.
  • Cat. no.:
    75793-612
  • Supplier No.:
    90-053

Specifications

About this item

Nicotinamide phosphoribosyltransferase (Nampt; pre-B cell colony-enhancing factor; PBEF; Visfatin) is an 52kDa adipokine secreted by adipose tissue and involved in the biosynthesis of nicotinamide adenine dinucleotide (NAD+). Two forms of Nampt exist, an intracellular form (iNampt) and an extracellular form (eNampt). While the function of iNampt as an essential and rate-limiting NAD+ biosynthetic enzyme is well established, the physiological role of eNampt is still a matter of debate. Nampt has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity. The protein is highly expressed in visceral fat and serum levels of the protein correlate with obesity.

  • Fusion-Tag: DDDDK Tag

This recombinant proteins is for research use only.

Customers Who Bought This Also Bought