To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 μg
- Tag sequence:His Tag
- Storage conditions:Working aliquots are stable for up to 3 months when stored at −20 °C.
- Biological activity:Shows adipogenic effects in stimulated differentiating 3T3-L1 cells.
- Protein synonyms:Pre-B Cell Colony Enhancing Factor 1|Nicotinamide Phosphoribosyltransferase|PBEF1
- Protein/peptide name:NAMPT
- Purity:>90% (SDS-PAGE)
- Sequence:1-491
- Formulation:Lyophilized, Salt-free.
- Cat. no.:75793-526
- Supplier No.:90-010
Specifications
About this item
Nicotinamide phosphoribosyltransferase (Nampt; pre-B cell colony-enhancing factor; PBEF; Visfatin) is an 52kDa adipokine secreted by adipose tissue and involved in the biosynthesis of nicotinamide adenine dinucleotide (NAD+). Two forms of Nampt exist, an intracellular form (iNampt) and an extracellular form (eNampt). While the function of iNampt as an essential and rate-limiting NAD+ biosynthetic enzyme is well established, the physiological role of eNampt is still a matter of debate. Nampt has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity. The protein is highly expressed in visceral fat and serum levels of the protein correlate with obesity.
- Fusion-Tag: His Tag
This recombinant proteins is for research use only.