To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:50 μg
- Tag sequence:His Tag
- Storage conditions:Working aliquots are stable for up to 3 months when stored at −20 °C.
- Biological activity:Shows adipogenic effects in stimulated differentiating 3T3-L1 cells.
- Protein synonyms:Pre-B Cell Colony Enhancing Factor 1|Nicotinamide Phosphoribosyltransferase|PBEF1
- Protein/peptide name:NAMPT
- Purity:>90% (SDS-PAGE)
- Sequence:1-491
- Formulation:Lyophilized from 0.2um-filtered solution in PBS, pH 7.2.
- Cat. no.:75793-524
- Supplier No.:90-009
Specifications
About this item
Nicotinamide phosphoribosyltransferase (Nampt; pre-B cell colony-enhancing factor; PBEF; Visfatin) is an adipokine that is localized to the bloodstream and has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity. The protein is highly expressed in visceral fat and serum levels of the protein correlate with obesity.
- Fusion-Tag: His Tag
This recombinant proteins is for research use only.