To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Species:Mouse
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:Alpha-type platelet-derived growth factor receptor|CD140a|Pdgfra|PDGF-R-alpha|Alpha platelet-derived growth factor receptor|Platelet-derived growth factor receptor alpha
- Protein/peptide name:PDGF R alpha
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:56.9 kD
- Sequence:Leu25-Glu524
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Cat. no.:75791-964
- Supplier No.:92-701
Specifications
About this item
Platelet-derived growth factor receptors (PDGFR) are cell surface tyrosine kinase receptors for members of the platelet-derived growth factor (PDGF) family. The PDGF family consists of PDGF-A, -B, -C and -D, which form either homo- or heterodimers (PDGF-AA, -AB, -BB, -CC, -DD). The four PDGFs are inactive in their monomeric forms. PDGFs bind to the protein tyrosine kinase receptors PDGF receptor- alpha and - beta . These two receptor isoforms dimerize upon binding the PDGF dimer, leading to three possible receptor combinations, namely - alpha alpha, - beta beta and - alpha beta . PDGFR alpha and PDGFR beta are members of the class III RTK family. Inappropriate PDGFR alpha and PDGFR beta signaling has been linked to a number of proliferative disorders.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.