To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-Fc tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:CD120b|Tumor necrosis factor receptor superfamily member 1B|TNFRSF1B|p75|p80 TNF-alpha receptor|TNF-R2|Tumor necrosis factor receptor type II|Tumor necrosis factor receptor 2|TNF-RII
- Protein/peptide name:TNF Receptor II
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:51.9 kD
- Sequence:Leu23-Asp257
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH4O. Please aliquot the reconstituted solution toPTTLGTIHSPEDVIKALADHHHHHHHHHE
- Cat. no.:75791-962
- Supplier No.:92-700
Specifications
About this item
Tumor necrosis factor receptor superfamily member 1B (TNFRSF1B) is a member of the tumor necrosis factor receptor superfamily. Human TNF RII contains four cysteinerich repeats in its ECD, which shares 58% and 56% amino acid sequence identity with the mouse and rat orthologs, respectively.TNF RII is expressed predominantly on cells of the hematopoietic lineage, such as T and natural killer cells, as well as on endothelial cells, microglia, astrocytes,neurons, oligodendrocytes, cardiac myocytes, thymocytes, and mesenchymal stem cells.TNF RII binds to the membranebound forms of TNF alpha and Lymphotoxin alpha/TNF beta ;soluble TNF is thought to signal predominately through TNF RI.Soluble TNF RII is believed to inhibit TNF biological activity by binding TNF thereby preventing it from activating membrane TNF receptors.
- Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.