Order Entry
ContactUsLinkComponent
Human Recombinant Albumin (from Serum)
Human Recombinant Albumin (from Serum)
Catalog #: 75791-942
Supplier:  Prosci
CAS Number:  
Human Recombinant Albumin (from Serum)
Catalog #: 75791-942
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Serum
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    HSA|serum albumin|HAS|serum of albumin
  • Protein/peptide name:
    Albumin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    66.5 kD
  • Sequence:
    Asp25-Leu609
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75791-942
  • Supplier No.:
    92-690

Specifications

About this item

Human serum albumin (HSA), the most abundant protein in human blood plasma, is essential for maintaining osmotic pressure. It is produced in the liver, consists of a single polypeptide chain, with 585 amino acids with 17 tyrosil residues and one tryptophan located in position 214. HSA is organized in three domains, I, II and III, each consisting of two subdomains, A and B. In the physiological states, HSA occurs in two forms – the non-modified HSA with a free thiol group of Cys-34, and the modified (oxidized) form (oHSA), also called human mercaptoalbumin (HMA) and human nonmercaptoalbumin (HNA), respectively. HMA and HNA are in equilibrium, depending on the redox state of Cys-34, and their ratio also varies depending on age and the diseased state. HSA functions primarily as a carrier protein for drugs, steroids, fatty acids, and thyroid hormones, and plays a role in stabilizing extracellular fluid volume.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought