To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Species:Mouse
- Size:0.05 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:Erythroid-Potentiating Activity|Metalloproteinase Inhibitor 1|Tissue Inhibitor of Metalloproteinases 1|CLGI|TIMP1|TIMP|Collagenase Inhibitor|Fibroblast collagenase Inhibitor|EPA|TIMP-1
- Protein/peptide name:TIMP1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:20.2 kD
- Sequence:Cys25-Arg205
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NaCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Cat. no.:75791-768
- Supplier No.:92-603
Specifications
About this item
Tissue Inhibitor of Metalloproteinases 1 (TIMP-1) complexes with metalloproteinases (such as collagenases) and irreversibly inactivates them by binding to their catalytic zinc cofactor. Also mediates erythropoiesis in vitro; but, unlike IL-3, it is species-specific, stimulating the growth and differentiation of only human and murine erythroid progenitors. Known to act on MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-10, MMP-11, MMP-12, MMP-13, and MMP-16. TIMP-1 does not act on MMP-14.
This recombinant protein can be used for biological assays. For research use only.