Order Entry
ContactUsLinkComponent
Mouse Recombinant IL-2 R alpha
Mouse Recombinant IL-2 R alpha
Catalog #: 75791-626
Supplier:  Prosci
CAS Number:  
Mouse Recombinant IL-2 R alpha
Catalog #: 75791-626
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    interleukin-2 receptor subunit alpha|alpha|IL-2RA|TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|interleukin 2 receptor|IDDM10|CD25 antigen|CD25|IL2RA|IL-2 R alpha
  • Protein/peptide name:
    IL-2 R alpha
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    55.5 kD
  • Sequence:
    Glu22-Lys236
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. no.:
    75791-626
  • Supplier No.:
    92-528

Specifications

About this item

The interleukin 2 receptor alpha (IL2RA), also called CD25, is a type I transmembrane protein that presents on activated T cells, activated B cells, some thymocytes, myeloid precursors, and oligodendrocytes. IL2RA is a member of cytokine receptors family that utilizes the common gamma chain subunit ( gamma c). IL2RA is expressed in most B-cell neoplasms, some acute nonlymphocytic leukemias, neuroblastomas, and tumor infiltrating lymphocytes. IL2RA associates with IL2RB (CD122) to form a heterodimer that can act as a high-affinity receptor for IL-2.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought