Order Entry
ContactUsLinkComponent
Human Recombinant HOXB4 (from E. coli)
Human Recombinant HOXB4 (from E. coli)
Catalog #: 75791-620
Supplier:  Prosci
CAS Number:  
Human Recombinant HOXB4 (from E. coli)
Catalog #: 75791-620
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    HOXB4|Homeobox protein Hox-2F|HOX-2F|Homeobox protein Hox-B4|HOX2F|Homeobox protein Hox-2.6
  • Protein/peptide name:
    HOXB4
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    29.8 kD
  • Sequence:
    Met1-Leu251
  • Formulation:
    Supplied as a 0.2 um filtered solution of 4mM HCl. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. no.:
    75791-620
  • Supplier No.:
    92-525

Specifications

About this item

Homeobox B4 (HOXB4) is encoded by the HOXB4 gene which is a member of the the class I homeobox (HOX) gene family and encodes a nuclear protein with a homeobox DNA-binding domain. These genes are master control regulators of developmental programs including embryonic and adult hematopoiesis. Multiple HOX genes, including HOXB4, are highly expressed in the hematopoietic stem cells (HSC) compartment. HOXB4 gene can act in opposite ways when expressed by different cells, promoting the proliferation of stem cells whilst activating the apoptotic pathway in some embryonic structures. The protein HOXB4, as a homeodomain transcription factor, has been shown to be an important regulator of stem cell renewal and hematopoiesis. Incellular or ectopic expression of HOXB4 expands hematopoietic stem and progenitor cells in vivo and in vitro, making it a potential candidate for therapeutic stem cell expansion.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought