Order Entry
ContactUsLinkComponent
Mouse Recombinant Carbonic anhydrase 14 (from E. coli)
Mouse Recombinant Carbonic anhydrase 14 (from E. coli)
Catalog #: 75791-616
Supplier:  Prosci
CAS Number:  
Mouse Recombinant Carbonic anhydrase 14 (from E. coli)
Catalog #: 75791-616
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Carbonic anhydrase 14
  • Enzyme synonyms:
    Carbonic Anhydrase 14, Carbonate Dehydratase XIV, Carbonic Anhydrase XIV, CA-XIV, CA14
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    32.3 kD
  • Sequence:
    Ala16-Met290
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris, 150 mM NaCl,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. no.:
    75791-616
  • Supplier No.:
    92-523

Specifications

About this item

Mouse Ca14,also known as Carbonic anhydrase 14,is a member of large family of zinc metalloenzymes .It could catalyze reversible hydration of carbon dioxide

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

The reaction is fundamental to many processes such as respiration, renal tubular acidification and bone resorption. Fifteen CA isoforms have been reported so far. They have different patterns of tissue-specific expression and physiologic roles. Some CAs may serve as markers for tumors and hypoxia. CA XIV is a polypeptide consisting of an extracellular N-terminal catalytic domain, a membrane-spanning segment and a short intracellular C- terminal segment with several potential phosphorylation sites. A subset of CAs lack CA activity due to point mutations but retain esterase function. CA14 is widely expressed in the central nervous system

Customers Who Bought This Also Bought