Order Entry
ContactUsLinkComponent
Mouse Recombinant IL1RA (from E. coli)
Mouse Recombinant IL1RA (from E. coli)
Catalog #: 75791-604
Supplier:  Prosci
CAS Number:  
Mouse Recombinant IL1RA (from E. coli)
Catalog #: 75791-604
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    IL-1F3|IL-1RN/IL-1ra/IRAP/IL1 inhibitor/Il1rn
  • Protein/peptide name:
    IL1RA
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.5 kD
  • Sequence:
    Arg27-Gln178
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. no.:
    75791-604
  • Supplier No.:
    92-517

Specifications

About this item

Interleukin-1 receptor antagonist protein (Il1rn), also known as IL-1ra, IRAP or IL1 inhibitor, is a member of the interleukin 1 cytokine family. This protein inhibits the activities of interleukin 1 alpha (IL1A) and interleukin 1 beta (IL1B), and modulates a variety of interleukin 1 related immune and inflammatory responses. The mouse Il1rn gene encodes a 178 amino acids (aa) protein with a 26 aa signal peptide. Mouse Il1rn protein shares 26% and 19% identity with its homologues IL-1 beta and IL-1 alpha, respectively. Il1rn can Inhibits the activity of interleukin-1 by binding to receptor IL1R1 and preventing its association with the coreceptor IL1RAP for signaling, but has no interleukin-1 like activity. Recently, an recombinant human Il1rn protein is used in the treatment of rheumatoid arthritis, an autoimmune disease in which IL-1 plays a key role.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought