Order Entry
ContactUsLinkComponent
Human Recombinant ECH1 (from E. coli)
Human Recombinant ECH1 (from E. coli)
Catalog #: 75791-580
Supplier:  Prosci
CAS Number:  
Human Recombinant ECH1 (from E. coli)
Catalog #: 75791-580
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    5)-Delta(2|mitochondrial|ECH1|4)-dienoyl-CoA isomerase|Delta(3|REF: C1008
  • Protein/peptide name:
    ECH1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    34.45 kD
  • Sequence:
    Thr34-Leu328
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM Tris,100mM NaCl,10% Glycerol,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. no.:
    75791-580
  • Supplier No.:
    92-505

Specifications

About this item

Human delta(3,5)-Delta(2,4)-dienoyl-CoA isomerase(ECH1) is a member of the hydratase/isomerase superfamily and contains a C-terminal peroxisomal targeting sequence and localizes to peroxisomes. ECH1 shows high sequence similarity to enoyl-CoA hydratases of several species, particularly within a conserved domain characteristic of these proteins. The rat orthologlocalizes to the matrix of both the peroxisome and mitochondria.It can isomerize 3-trans, 5-cis-dienoyl-CoA to 2-trans,4-trans-dienoyl-CoA, indicating that it is a delta3,5-delta2,4-dienoyl-CoA isomerase. ECH1 plays an important role in the auxiliary step of the fatty acid beta-oxidation pathway.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought