To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Species:Mouse
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:Osteoprotegerin ligand|OPGL|ODF|TRANCE|Receptor activator of nuclear factor kappa-B ligand|CD254|TNF-related activation-induced cytokine|RANKL|Osteoclast differentiation factor|RANK L|Tumor necrosis factor ligand superfamily member 11|Tnfsf11
- Protein/peptide name:TRANCE
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:28.5 kD
- Sequence:Ala73-Asp316
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Cat. no.:75791-554
- Supplier No.:92-492
Specifications
About this item
Mouse tumor necrosis factor ligand superfamily member 11(Tnfsf11) is a member of the tumor necrosis factor (TNF) cytokine family. Tnfsf11 is widely expressed in cells including T cells and T cell rich organs, such as thymus and lymph nodes. This cytokine can bind to TNFRSF11B/OPG andTNFRSF11A/RANK. Tnfsf11 is involved in a number of fundamental biological processes such as acting as regulator of interactions between T-cells and dendritic cells, the regulation of the T-cell-dependent immune response and enhancing bone-resorption in humoral hypercalcemia of malignancy. It augments the ability of dendritic cells to stimulate naive T-cell proliferation.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.