Order Entry
ContactUsLinkComponent
Human Recombinant IL-2 R alpha (from Cells)
Human Recombinant IL-2 R alpha (from Cells)
Catalog #: 75791-522
Supplier:  Prosci
CAS Number:  
Human Recombinant IL-2 R alpha (from Cells)
Catalog #: 75791-522
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    IL-2RA|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|TAC antigen|Interleukin-2 receptor subunit alpha|CD25
  • Protein/peptide name:
    IL-2 R alpha
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    48.9 kD
  • Sequence:
    Glu22-Cys213
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. no.:
    75791-522
  • Supplier No.:
    92-476

Specifications

About this item

Interleukin-2 receptor subunit alpha (IL2RA) is a single-pass type I membrane protein, contains 2 Sushi (CCP/SCR) domains. The interleukin 2 (IL2) receptor alpha (IL2RA) and beta (IL2RB) chains, together with the common gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric alpha chains (IL2RA) result in low-affinity receptor, while homodimeric beta (IL2RB) chains produce a medium-affinity receptor. Normally an integral-membrane protein, soluble IL2RA has been isolated and determined to result from extracellular proteolyisis.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought