To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Enzyme type:Recombinant
- Species:Mouse
- Size:0.05 mg
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:CNDP dipeptidase 2
- Enzyme synonyms:CNDP2/CNDP dipeptidase 2/Cytosolic non-specific dipeptidase/Glutamate carboxypeptidase-like protein 1/Cn2, CNDP2, CPGL
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:53.8 kD
- Sequence:Met1-Asn475
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
- Cat. no.:75791-402
- Supplier No.:92-416
Specifications
About this item
Mouse cytosolic non-specific dipeptidase(CNDP2) is a cytoplasm protein which belongs to the peptidase M20A family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
CNDP2 has 2 Isoform: Isoform 1 is ubiquitously expressed with higher levels in kidney and liver (at protein level). Isoform 2 is expressed in fetal tissues, it is only expressed in adult liver and placental tissues. CNDP2 hydrolyzes a variety of dipeptides including L-carnosine and has a strong preference for Cys-Gly. It is a cytosolic enzyme that can hydrolyze carnosine to yield l-histidine and beta-alanine. CNDP2 is highly expressed in the histaminergic neurons in the tuberomammillary nucleus, implying that it may supply histidine to histaminergic neurons for histamine synthesis. It may play a role as tumor suppressor in hepatocellular carcinoma (HCC) cells.