Order Entry
ContactUsLinkComponent
Human Recombinant IL12
Human Recombinant IL12
Catalog #: 75791-312
Supplier:  Prosci
CAS Number:  
Human Recombinant IL12
Catalog #: 75791-312
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    NK cell|Interleukin-12 subunit alpha|Cytotoxic lymphocyte maturation factor 35 kDa subunit|IL-12A|IL-12 subunit p35|IL12A|CLMF p35|NKSF1 stimulatory factor chain 1
  • Protein/peptide name:
    IL12
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    22.5 and 34.7 kD
  • Sequence:
    Arg23-Ser219&Ile23-Ser328
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75791-312
  • Supplier No.:
    92-363

Specifications

About this item

IL-12 is involved in the differentiation of naive T cells into Th1 cells. It is known as a T cell-stimulating factor, which can stimulate the growth and function of T cells. It stimulates the production of interferon-gamma (IFN- gamma ) and tumor necrosis factor-alpha (TNF- alpha) from T cells and natural killer (NK) cells, and reduces IL-4 mediated suppression of IFN- gamma . T cells that produce IL-12 have a coreceptor, CD30, which is associated with IL-12 activity.IL-12 plays an important role in the activities of natural killer cells and T lymphocytes. IL-12 mediates enhancement of the cytotoxic activity of NK cells and CD8+ cytotoxic T lymphocytes.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought