To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:N-GST tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:Cytochrome b-c1 complex subunit 6|Complex III subunit VIII|Complex III subunit 6|Mitochondrial hinge protein|Ubiquinol-cytochrome c reductase complex 11 kDa protein|mitochondrial|Cytochrome c1 non-heme 11 kDa protein
- Protein/peptide name:UQCRH
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:37.5 kD
- Sequence:Met1-Lys91
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75791-150
- Supplier No.:92-278
Specifications
About this item
Cytochrome b-c1 complex subunit 6, mitochondrial (UQCRH) belongs to the UQCRH/QCR6 family, it is a subunit of the respiratory chain protein Ubiquinol Cytochrome c Reductase. This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. UQCRH may mediate formation of the complex between cytochromes c and c1. It may mediate formation of the complex between cytochromes c and c1.
- Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.