Order Entry
ContactUsLinkComponent
Human Recombinant TRIM5 (from E. coli)
Human Recombinant TRIM5 (from E. coli)
Catalog #: 75791-102
Supplier:  Prosci
CAS Number:  
Human Recombinant TRIM5 (from E. coli)
Catalog #: 75791-102
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Tripartite motif-containing protein 5|RNF88|TRIM5|RING finger protein 88
  • Protein/peptide name:
    TRIM5
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    30.8 kD
  • Sequence:
    Met1-Gln248
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75791-102
  • Supplier No.:
    92-254

Specifications

About this item

Tripartite motif-containing Motif 5 is a protein that in humans is encoded by the TRIM5 gene.It is a 493 amino acids protein that belongs to the TRIM/RBCC family.It contains 1 B box-type zinc finger, 1 B30.2/SPRY domain and 1 RING-type zinc finger. TRIM5 present in the cytoplasm recognizes motifs within the capsid proteins and interferes with the uncoating process, therefore preventing successful reverse transcription and transport to the nucleus of the viral genome. The exact mechanism of action has not been shown conclusively, but capsid protein from restricted viruses is removed by proteasome-dependent degradation

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought