Order Entry
ContactUsLinkComponent
Human Recombinant FKBP52 (from E. coli)
Human Recombinant FKBP52 (from E. coli)
Catalog #: 75790-990
Supplier:  Prosci
CAS Number:  
Human Recombinant FKBP52 (from E. coli)
Catalog #: 75790-990
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    59kDa immunophilin|HSP-binding immunophilin|FK506-binding protein 4|Peptidyl-prolyl cis-trans isomerase FKBP4|FKBP59|Rotamase|51kDa FK506-binding protein|immunophilin FKBP52|FKBP4
  • Protein/peptide name:
    FKBP52
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    52.8 kD
  • Sequence:
    Met1-Ala459
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM Tris,10%glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-990
  • Supplier No.:
    92-198

Specifications

About this item

FKBP4 act as a regulator of microtubule dynamics by inhibiting MAPT/TAU ability to promote microtubule assembly. FKBP4 may play a role in the intracellular trafficking of heterooligomeric forms of steroid hormone receptors between cytoplasm and nuclear compartments, it also may have a protective role against oxidative stress in mitochondria. The isomerase activity controls neuronal growth cones via regulation of TRPC1 channel opening.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought