To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:PDN-21|CCP|CALC1|Calcitonin|Katacalcin|Calcitonin Carboxyl-Terminal Peptide|CALCA
- Protein/peptide name:Calcitonin
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:10.5 kD
- Sequence:Tyr58-Asn141
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,50% Glycerol,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-956
- Supplier No.:92-181
Specifications
About this item
Calcitonin is a secreted protein which belongs to the calcitonin family. Calcitonin is cleaved into the following two chains: Calcitonin and Katacalcin. Katacalcin is a potent plasma calcium-lowering peptide. Calcitonin is a 32-amino acid linear polypeptide hormone. Calcitonin acts to reduce blood calcium (Ca2+), opposing the effects of parathyroid hormone (PTH). Its importance in humans has not been as well established as its importance in other animals, as its function is usually not significant in the regulation of normal calcium homeostasis. Calcitonin causes a rapid but short-lived drop in the level of calcium and phosphate in blood by promoting the incorporation of those ions in the bones.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.